siRNA Details
Gene Symbol: Taf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037310.1 | NP_001032387.1 | 3 | 1298 | ||
NM_184048.2 | NP_908937.2 | 3 | 819 | |||
GenBank mRNA | AY325235.1 | - | 2 | 655 | ||
AY359819.1 | - | 1 | 615 | |||
BC091109.1 | - | 3 | 808 | 7322630 | ||
FQ213120.1 | - | 3 | 802 | |||
FQ215462.1 | - | 3 | 813 | |||
FQ229736.1 | - | 3 | 812 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
