siRNA Details
Gene Symbol: SLC7A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012244.3 | NP_036376.2 | 2 | 943 | ||
GenBank mRNA | AB037669.1 | - | 2 | 906 | ||
AF135828.1 | - | 2 | 418 | |||
AF171669.1 | - | 2 | 420 | |||
AK223610.1 | - | 2 | 942 | |||
AK313465.1 | - | 2 | 197 | |||
AL365342.1 | - | 2 | 928 | |||
AL365343.2 | - | 2 | 447 | |||
AL365414.1 | - | 2 | 447 | |||
BC052250.1 | - | 2 | 226 | 6260264 | ||
Y18483.1 | - | 2 | 927 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
