siRNA Details
Gene Symbol: Klra5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198746.1 | NP_942041.1 | 4 | 482 | ||
XM_008763341.1 | XP_008761563.1 | 4 | 638 | |||
XM_008763342.1 | XP_008761564.1 | 4 | 458 | |||
XM_008763343.1 | XP_008761565.1 | 4 | 368 | |||
XM_008763344.1 | XP_008761566.1 | 669 | ||||
GenBank mRNA | U56863.1 | - | 4 | 482 |
Gene Symbol: LOC100911272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763440.1 | XP_008761662.1 | 4 | 1195 |
Gene Symbol: Ly49i7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009500.1 | NP_001009500.1 | 3 | 339 | ||
XM_008763320.1 | XP_008761542.1 | 4 | 1436 | |||
GenBank mRNA | AY659934.1 | - | 3 | 339 |

More Information
Other Species Targeted

Gene Ontology Categories:
