siRNA Details
Gene Symbol: Srr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198757.2 | NP_942052.1 | 4 | 533 | ||
XM_008768010.1 | XP_008766232.1 | 3 | 448 | |||
XM_008768011.1 | XP_008766233.1 | 3 | 446 | |||
XM_008768012.1 | XP_008766234.1 | 3 | 329 | |||
XM_008768013.1 | XP_008766235.1 | 3 | 329 | |||
XM_008768014.1 | XP_008766236.1 | 3 | 283 | |||
XM_008768015.1 | XP_008766237.1 | 3 | 351 | |||
GenBank mRNA | AB106282.1 | - | 3 | 303 | ||
BC082014.1 | - | 4 | 533 | 7132082 |
