siRNA Details
Gene Symbol: Gpcpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198779.1 | NP_942074.1 | 4 | 269 | ||
XM_008762183.1 | XP_008760405.1 | 2 | 2132 | |||
XM_008762184.1 | XP_008760406.1 | 2 | 2131 | |||
XM_008762185.1 | XP_008760407.1 | 4 | 330 | |||
XM_008762186.1 | XP_008760408.1 | 4 | 333 | |||
GenBank mRNA | AY233980.1 | - | 4 | 269 | ||
FQ217815.1 | - | 4 | 287 | |||
FQ217938.1 | - | 4 | 287 |
