siRNA Details
Gene Symbol: Dgki | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198782.2 | NP_942077.2 | 6 | 1010 | ||
XM_006236333.2 | XP_006236395.1 | 6 | 832 | |||
GenBank mRNA | AB058962.1 | - | 6 | 1010 | ||
AB058963.1 | - | 6 | 1010 | |||
AB058964.1 | - | 6 | 1010 |

More Information
Other Species Targeted

Gene Ontology Categories:
