siRNA Details
Gene Symbol: Tubb4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199094.2 | NP_954525.1 | 5 | 1537 | ||
XM_006233582.2 | XP_006233644.1 | 5 | 1550 | |||
GenBank mRNA | BC060597.1 | - | 5 | 1541 | 6889737 | |
FQ213825.1 | - | 5 | 1536 | |||
FQ215022.1 | - | 5 | 1536 | |||
FQ227178.1 | - | 5 | 1534 | |||
FQ228801.1 | - | 5 | 1534 |

More Information
Other Species Targeted

Gene Ontology Categories:
