siRNA Details
Gene Symbol: Fam198b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199105.1 | NP_954536.1 | 5 | 1944 | ||
XM_006232514.2 | XP_006232576.1 | 6 | 2275 | |||
GenBank mRNA | BC061747.1 | - | 5 | 1944 | 5599685 |
