siRNA Details
Gene Symbol: CBX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127321.1 | NP_001120793.1 | 2, 3 | 255 | ||
NM_001127322.1 | NP_001120794.1 | 2, 3 | 453 | |||
NM_012117.2 | NP_036249.1 | 2, 3 | 301 | |||
GenBank mRNA | AK313506.1 | - | 2, 3 | 262 | ||
BC006821.1 | - | 2, 3 | 203 | 3448801 | ||
BP236173.1 | - | 2, 3 | 255 | |||
BP336150.1 | - | 2, 3 | 301 | |||
CR457418.1 | - | 1, 2 | 135 | |||
DT217255.1 | - | 2, 3 | 453 | |||
EU794599.1 | - | 2, 3 | 270 | |||
L07515.1 | - | 2, 3 | 270 | |||
S62077.1 | - | 2, 3 | 269 | |||
U26311.1 | - | 1, 2 | 133 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001076789, NM_007626, NM_001110216, X99641, AK032975, and others.
Gene Ontology Categories:
