siRNA Details
Gene Symbol: Popdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199113.1 | NP_954544.1 | 1 | 563 | ||
XM_008768733.1 | XP_008766955.1 | 1 | 2570 | |||
XM_008768734.1 | XP_008766956.1 | 1 | 2570 | |||
GenBank mRNA | AY490241.1 | - | 1 | 526 | ||
BC061876.1 | - | 1 | 563 | 5599130 |
