siRNA Details
Gene Symbol: Tmbim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199116.2 | NP_954547.1 | 2 | 181 | ||
GenBank mRNA | BC060596.1 | - | 2 | 182 | 6889044 | |
FQ217672.1 | - | 2 | 124 | |||
FQ228009.1 | - | 2 | 178 | |||
FQ230396.1 | - | 2 | 146 | |||
FQ233689.1 | - | 2 | 175 |
