siRNA Details
Gene Symbol: Abcc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199377.1 | NP_955409.1 | 5, 6 | 734 | ||
XM_006255241.2 | XP_006255303.1 | 5, 6 | 875 | |||
XM_006255242.2 | XP_006255304.1 | 5, 6 | 876 | |||
XM_008772375.1 | XP_008770597.1 | 5, 6 | 877 | |||
XM_008772376.1 | XP_008770598.1 | 5, 6 | 874 | |||
XM_008772377.1 | XP_008770599.1 | 5, 6 | 878 | |||
XM_008772378.1 | XP_008770600.1 | 5, 6 | 878 | |||
GenBank mRNA | AY188179.1 | - | 5, 6 | 734 |
