siRNA Details
Gene Symbol: Abcc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199377.1 | NP_955409.1 | 21 | 2800 | ||
XM_006255241.2 | XP_006255303.1 | 21 | 2941 | |||
XM_006255242.2 | XP_006255304.1 | 20 | 2738 | |||
XM_008772375.1 | XP_008770597.1 | 21 | 2964 | |||
XM_008772376.1 | XP_008770598.1 | 21 | 2961 | |||
XM_008772377.1 | XP_008770599.1 | 21 | 2965 | |||
XM_008772378.1 | XP_008770600.1 | 21 | 2965 | |||
GenBank mRNA | AY188179.1 | - | 21 | 2800 | ||
AY188180.1 | - | 20 | 2563 |
