siRNA Details
Gene Symbol: Tgfbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012775.2 | NP_036907.2 | 323 | |||
XM_006238030.2 | XP_006238092.2 | 325 | ||||
GenBank mRNA | BC087035.1 | - | 265 | 7098473 | ||
CB804748.1 | - | 367 | ||||
L26110.1 | - | 280 |
Gene Symbol: LOC103690035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238153.2 | XP_006238215.2 | 1 | 1608 | ||
XM_008763706.1 | XP_008761928.1 | 1 | 1608 |
