siRNA Details
Gene Symbol: Bmp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012827.2 | NP_036959.2 | 235 | |||
XM_006251746.2 | XP_006251808.1 | 997 | ||||
XM_006251747.2 | XP_006251809.1 | 547 | ||||
XM_006251748.2 | XP_006251810.1 | 3 | 529 | |||
XM_006251749.2 | XP_006251811.1 | 271 | ||||
XM_006251751.2 | XP_006251813.1 | 248 | ||||
XM_008770501.1 | XP_008768723.1 | 361 | ||||
GenBank mRNA | AY184241.1 | - | 1 | 97 | ||
BC078901.1 | - | 235 | 7121295 | |||
Z22607.1 | - | 3 | 497 |

More Information
Other Species Targeted

Gene Ontology Categories:
