siRNA Details
Gene Symbol: Egf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012842.1 | NP_036974.1 | 908 | |||
GenBank mRNA | BC062030.1 | - | 893 | 5623849 | ||
JX149564.1 | - | 752 | ||||
JX437943.1 | - | 752 | ||||
U04842.1 | - | 908 |
Gene Symbol: LOC100910178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760814.1 | XP_008759036.1 | 3, 4 | 919 | ||
XM_008760815.1 | XP_008759037.1 | 3, 4 | 918 | |||
XM_008760816.1 | XP_008759038.1 | 3, 4 | 679 |
