siRNA Details
Gene Symbol: Ghrhr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012850.1 | NP_036982.1 | 2 | 110 | ||
XM_008762872.1 | XP_008761094.1 | 2 | 1473 | |||
GenBank mRNA | AF122055.1 | - | 2 | 83 | ||
L01407.1 | - | 2 | 110 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
