siRNA Details
Gene Symbol: Ghrhr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012850.1 | NP_036982.1 | 13 | 1505 | ||
GenBank mRNA | L01407.1 | - | 13 | 1505 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ghrhr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012850.1 | NP_036982.1 | 13 | 1505 | ||
GenBank mRNA | L01407.1 | - | 13 | 1505 |
Set Membership: |
![]() |