siRNA Details
Gene Symbol: Spp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012881.2 | NP_037013.2 | 6 | 413 | ||
XM_008769996.1 | XP_008768218.1 | 6 | 486 | |||
GenBank mRNA | AB001382.1 | - | 5 | 307 | ||
BC078874.1 | - | 6 | 393 | 7110677 | ||
CV102664.1 | - | 6 | 413 | |||
FQ213473.1 | - | 6 | 402 | |||
FQ222192.1 | - | 6 | 429 | |||
M14656.1 | - | 6 | 386 | |||
M99252.1 | - | 6 | 422 |
