siRNA Details
Gene Symbol: Tmpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012887.1 | NP_037019.1 | 2 | 596 | ||
XM_008765218.1 | XP_008763440.1 | 2 | 635 | |||
XM_008765219.1 | XP_008763441.1 | 2 | 635 | |||
XM_008765220.1 | XP_008763442.1 | 2 | 635 | |||
GenBank mRNA | U18314.1 | - | 2 | 596 |
