siRNA Details
Gene Symbol: Alad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012899.2 | NP_037031.1 | 2 | 259 | ||
XM_006238234.2 | XP_006238296.1 | 2 | 852 | |||
XM_006238235.1 | XP_006238297.1 | 2 | 349 | |||
GenBank mRNA | BC061806.1 | - | 2 | 259 | 5598340 | |
FQ210250.1 | - | 2 | 204 | |||
FQ213585.1 | - | 2 | 401 | |||
FQ218546.1 | - | 2 | 204 | |||
FQ219571.1 | - | 2 | 204 | |||
FQ231154.1 | - | 2 | 235 | |||
FQ232546.1 | - | 2 | 235 | |||
FQ233724.1 | - | 2 | 285 | |||
FQ234245.1 | - | 2 | 235 | |||
FQ234573.1 | - | 2 | 283 | |||
X04959.1 | - | 1 | 141 |
