siRNA Details
Gene Symbol: Ambp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012901.1 | NP_037033.1 | 7 | 671 | ||
XM_008763759.1 | XP_008761981.1 | 7 | 757 | |||
GenBank mRNA | BC088166.1 | - | 7 | 656 | 7368246 | |
FQ209788.1 | - | 4 | 239 | |||
FQ209831.1 | - | 4 | 239 | |||
FQ210037.1 | - | 4 | 239 | |||
FQ210364.1 | - | 4 | 239 | |||
FQ210824.1 | - | 4 | 239 | |||
FQ219366.1 | - | 4 | 239 | |||
FQ219628.1 | - | 4 | 240 | |||
FQ219767.1 | - | 4 | 239 | |||
S87544.1 | - | 7 | 671 |
