siRNA Details
Gene Symbol: Cacna2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110847.1 | NP_001104317.1 | 6 | 450 | ||
NM_001110848.1 | NP_001104318.1 | 6 | 438 | |||
NM_012919.3 | NP_037051.2 | 6 | 723 | |||
XM_006235981.2 | XP_006236043.1 | 6 | 713 | |||
XM_006235982.2 | XP_006236044.1 | 6 | 713 | |||
XM_006235983.2 | XP_006236045.1 | 6 | 713 | |||
GenBank mRNA | AF286488.1 | - | 6 | 724 | ||
AF400662.1 | - | 6 | 450 | |||
AF486276.1 | - | 6 | 438 | |||
M86621.1 | - | 6 | 592 |
