siRNA Details
Gene Symbol: Cd44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012924.2 | NP_037056.2 | 19 | 3100 | ||
XM_006234627.2 | XP_006234689.1 | 16 | 2876 | |||
XM_006234629.2 | XP_006234691.1 | 14 | 2642 | |||
XM_006234631.2 | XP_006234693.1 | 9 | 1974 | |||
XM_008762055.1 | XP_008760277.1 | 15 | 2785 | |||
XM_008762056.1 | XP_008760278.1 | 14 | 2645 | |||
XM_008762057.1 | XP_008760279.1 | 13 | 2553 | |||
XM_008762058.1 | XP_008760280.1 | 12 | 2349 | |||
XM_008762059.1 | XP_008760281.1 | 19 | 3240 | |||
GenBank mRNA | BC061531.1 | - | 9 | 1857 | ||
BC085703.1 | - | 9 | 1884 | |||
BC107949.1 | - | 9 | 1840 | |||
M61874.1 | - | 12 | 2269 | |||
M61875.1 | - | 9 | 1852 | |||
U96138.1 | - | 19 | 3100 |
