siRNA Details
Gene Symbol: Gabrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012956.1 | NP_037088.1 | 2 | 180 | ||
GenBank mRNA | X15466.1 | - | 2 | 180 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008069, BC157920, BC130258.
Gene Ontology Categories:
