siRNA Details
Gene Symbol: Gabrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246127.2 | XP_006246189.1 | 10 | 1919 | ||
XM_008767635.1 | XP_008765857.1 | 11 | 2019 | |||
GenBank mRNA | X15467.1 | - | 9 | 1534 |
