siRNA Details
Gene Symbol: Kcna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012970.3 | NP_037102.1 | 3 | 2096 | ||
XM_006233132.2 | XP_006233194.1 | 3 | 2682 | |||
XM_006233133.2 | XP_006233195.1 | 3 | 2264 | |||
XM_006233134.2 | XP_006233196.1 | 3 | 2247 | |||
XM_006233135.2 | XP_006233197.1 | 3 | 2051 | |||
XM_008761371.1 | XP_008759593.1 | 3 | 2292 | |||
GenBank mRNA | J04731.1 | - | 3 | 2096 | ||
M74449.1 | - | 1 | 1565 | |||
X16003.1 | - | 3 | 2077 |
