siRNA Details
Gene Symbol: Kcna4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012971.2 | NP_037103.1 | 1 | 1280 | ||
XM_006234673.2 | XP_006234735.1 | 2 | 2476 | |||
XM_006234674.2 | XP_006234736.1 | 2 | 2328 | |||
GenBank mRNA | M32867.1 | - | 1 | 1280 | ||
X16002.1 | - | 1 | 1694 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_021275, AK036112, BC109014.
Gene Ontology Categories:
