siRNA Details
Gene Symbol: Lhcgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012978.1 | NP_037110.1 | 4, 5 | 449 | ||
GenBank mRNA | M26199.1 | - | 4, 5 | 449 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013582, AK030325, AK138461, M81310, AK054331.
Gene Ontology Categories:
