siRNA Details
Gene Symbol: P2rx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142367.2 | NP_001135839.1 | 12 | 1929 | ||
NM_001285415.1 | NP_001272344.1 | 11 | 1929 | |||
NM_012997.3 | NP_037129.1 | 12 | 2010 | |||
GenBank mRNA | BC061742.1 | - | 12 | 2002 | 5599053 |
