siRNA Details
Gene Symbol: Pam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013000.2 | NP_037132.2 | 1 | 271 | ||
XM_006245594.2 | XP_006245656.1 | 1 | 484 | |||
XM_006245595.2 | XP_006245657.1 | 1 | 484 | |||
XM_006245596.2 | XP_006245658.1 | 1 | 484 | |||
XM_006245597.2 | XP_006245659.1 | 1 | 484 | |||
XM_006245598.2 | XP_006245660.1 | 1 | 483 | |||
XM_006245599.2 | XP_006245661.1 | 1 | 483 | |||
XM_008767360.1 | XP_008765582.1 | 1 | 484 | |||
GenBank mRNA | M25719.1 | - | 1 | 387 | ||
M25732.1 | - | 1 | 271 | |||
M82845.1 | - | 1 | 142 | |||
X59689.1 | - | 1 | 92 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013626, U79523, AK051649, AK049423.
Gene Ontology Categories:
