siRNA Details
Gene Symbol: Phex | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013004.1 | NP_037136.1 | 2 | 634 | ||
GenBank mRNA | AJ001637.1 | - | 2 | 634 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: U75646.
Gene Ontology Categories:
