siRNA Details
Gene Symbol: Pik3r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013005.1 | NP_037137.1 | 11, 12 | 1576 | ||
XM_006231868.2 | XP_006231930.1 | 6, 7 | 1762 | |||
XM_008760659.1 | XP_008758881.1 | 6, 7 | 3212 | |||
GenBank mRNA | D64045.1 | - | 11, 12 | 1576 | ||
D64048.1 | - | 6, 7 | 766 | |||
D78486.1 | - | 6, 7 | 676 | |||
U50412.1 | - | 6, 7 | 770 |
