siRNA Details
Gene Symbol: Sirpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013016.2 | NP_037148.2 | 3 | 434 | ||
XM_006234951.2 | XP_006235013.1 | 2 | 732 | |||
XM_006234952.2 | XP_006235014.1 | 3 | 1094 | |||
XM_006234953.2 | XP_006235015.1 | 3 | 1276 | |||
GenBank mRNA | AF055065.1 | - | 3 | 298 | ||
BC099773.1 | - | 3 | 370 | 7932298 | ||
D38468.1 | - | 2 | 489 | |||
D85183.1 | - | 2 | 246 | |||
U62328.1 | - | 3 | 434 |
