siRNA Details
Gene Symbol: Sirpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013016.2 | NP_037148.2 | 3 | 471 | ||
XM_006234951.2 | XP_006235013.1 | 2 | 769 | |||
XM_006234952.2 | XP_006235014.1 | 3 | 1131 | |||
XM_006234953.2 | XP_006235015.1 | 3 | 1313 | |||
GenBank mRNA | AF055065.1 | - | 3 | 335 | ||
BC099773.1 | - | 3 | 407 | 7932298 | ||
D38468.1 | - | 2 | 526 | |||
D85183.1 | - | 2 | 283 | |||
U62328.1 | - | 3 | 471 |
