siRNA Details
Gene Symbol: SLC7A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012244.3 | NP_036376.2 | 2 | 1067 | ||
GenBank mRNA | AB037669.1 | - | 2 | 1030 | ||
AF087911.1 | - | 1 | 100 | |||
AF135828.1 | - | 2 | 542 | |||
AF171669.1 | - | 2 | 544 | |||
AK223610.1 | - | 2 | 1066 | |||
AK313465.1 | - | 2 | 321 | |||
AL365342.1 | - | 2 | 1052 | |||
AL365343.2 | - | 2 | 571 | |||
AL365414.1 | - | 2 | 571 | |||
BC052250.1 | - | 2 | 350 | 6260264 | ||
Y18483.1 | - | 2 | 1051 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
