siRNA Details
Gene Symbol: Slc5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013033.2 | NP_037165.1 | 8 | 926 | ||
GenBank mRNA | AB000729.1 | - | 8 | 952 | ||
BC081827.1 | - | 8 | 958 | 7129017 | ||
D16101.1 | - | 8 | 926 | |||
U03120.1 | - | 8 | 884 |
