siRNA Details
Gene Symbol: Slc6a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013034.4 | NP_037166.2 | 3, 4 | 802 | ||
XM_008767950.1 | XP_008766172.1 | 4, 5 | 858 | |||
GenBank mRNA | M79450.1 | - | 5, 6 | 926 | ||
X63253.1 | - | 3, 4 | 810 | |||
X63995.1 | - | 4, 5 | 855 | |||
Y11024.1 | - | 3, 4 | 695 |
