siRNA Details
Gene Symbol: Slc6a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013034.4 | NP_037166.2 | 13 | 2350 | ||
XM_008767950.1 | XP_008766172.1 | 14 | 2406 | |||
GenBank mRNA | M79450.1 | - | 15 | 2457 | ||
X63253.1 | - | 13 | 2358 | |||
X63995.1 | - | 14 | 2403 |
