siRNA Details
Gene Symbol: Tsc22d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109912.1 | NP_001103382.1 | 3 | 3254 | ||
NM_013043.2 | NP_037175.1 | 3 | 382 | |||
XM_006252339.1 | XP_006252401.1 | 4 | 3126 | |||
GenBank mRNA | BC059146.1 | - | 3 | 393 | ||
BC127547.1 | - | 3 | 374 | |||
CB771530.1 | - | 3 | 382 | |||
L25785.1 | - | 3 | 375 |
Gene Symbol: Tsc22d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191960.1 | NP_001178889.1 | 4 | 2068 | ||
XM_006232386.2 | XP_006232448.1 | 3 | 2897 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001081229, AK007760, AK156074, BC058221, EU004151, and others.
Gene Ontology Categories:
