siRNA Details
Gene Symbol: Id2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013060.3 | NP_037192.1 | 2 | 440 | ||
GenBank mRNA | BC086391.1 | - | 435 | 7303450 | ||
D10863.1 | - | 2 | 440 | |||
FQ212969.1 | - | 458 | ||||
FQ213471.1 | - | 458 | ||||
FQ213662.1 | - | 458 | ||||
FQ220574.1 | - | 2 | 458 | |||
FQ227397.1 | - | 2 | 458 |
