siRNA Details
Gene Symbol: ANGPTL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF125175.1 | - | 3 | 1044 | ||
AK075026.1 | - | 3 | 325 | |||
AK312876.1 | - | 4 | 1442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
