siRNA Details
Gene Symbol: Tub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013077.1 | NP_037209.1 | 3, 4 | 251 | ||
XM_008759690.1 | XP_008757912.1 | 3, 4 | 527 | |||
XM_008759691.1 | XP_008757913.1 | 3, 4 | 528 | |||
GenBank mRNA | AB011544.1 | - | 3, 4 | 251 |
