siRNA Details
Gene Symbol: Tub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013077.1 | NP_037209.1 | 7 | 729 | ||
XM_008759690.1 | XP_008757912.1 | 7 | 1005 | |||
XM_008759691.1 | XP_008757913.1 | 6 | 838 | |||
GenBank mRNA | AB011544.1 | - | 7 | 729 |

More Information
Other Species Targeted

Gene Ontology Categories:
