siRNA Details
Gene Symbol: Scn3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013119.1 | NP_037251.1 | 18, 19 | 3788 | ||
XM_008761914.1 | XP_008760136.1 | 17, 18 | 3687 | |||
XM_008761915.1 | XP_008760137.1 | 18, 19 | 3690 | |||
XM_008761916.1 | XP_008760138.1 | 17, 18 | 3688 | |||
XM_008761917.1 | XP_008760139.1 | 17, 18 | 3687 | |||
XM_008761918.1 | XP_008760140.1 | 17, 18 | 3600 | |||
XM_008761919.1 | XP_008760141.1 | 17, 18 | 3591 | |||
XM_008761920.1 | XP_008760142.1 | 17, 18 | 3591 | |||
GenBank mRNA | AM905325.1 | - | 4, 5 | 554 | ||
Y00766.1 | - | 18, 19 | 3788 |
