siRNA Details
Gene Symbol: Slc11a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013173.2 | NP_037305.2 | 3 | 185 | ||
XM_006242309.2 | XP_006242371.1 | 3 | 310 | |||
XM_006242310.2 | XP_006242372.1 | 3 | 311 | |||
XM_006242311.2 | XP_006242373.1 | 3 | 220 | |||
XM_006242312.2 | XP_006242374.1 | 3 | 215 | |||
GenBank mRNA | AF008439.1 | - | 3 | 196 | ||
AF029757.1 | - | 2 | 92 | |||
BC097338.1 | - | 2 | 112 | 7460406 |
