siRNA Details
Gene Symbol: Kcnj8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017099.4 | NP_058795.3 | 3 | 951 | ||
GenBank mRNA | AB043636.1 | - | 2 | 781 | ||
AB043637.1 | - | 2 | 781 | |||
BC078863.1 | - | 934 | 7128284 | |||
D42145.1 | - | 3 | 954 |
