siRNA Details
Gene Symbol: Grn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145842.1 | NP_001139314.1 | 2 | 173 | ||
NM_017113.2 | NP_058809.2 | 2 | 173 | |||
XM_008767981.1 | XP_008766203.1 | 2 | 254 | |||
GenBank mRNA | BC072469.1 | - | 2 | 167 | 7098196 | |
CB731349.1 | - | 2 | 204 | |||
CK365572.1 | - | 2 | 166 | |||
M97750.1 | - | 1 | 131 | |||
X62322.1 | - | 2 | 147 |

More Information
Other Species Targeted

Gene Ontology Categories:
