siRNA Details
Gene Symbol: Capn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017117.1 | NP_058813.1 | 1 | 563 | ||
XM_008762080.1 | XP_008760302.1 | 1 | 1195 | |||
XM_008762081.1 | XP_008760303.1 | 1 | 1195 | |||
GenBank mRNA | J05121.1 | - | 1 | 563 |
