siRNA Details
Gene Symbol: Capn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017117.1 | NP_058813.1 | 14 | 2059 | ||
XM_006234728.2 | XP_006234790.1 | 14 | 1793 | |||
XM_006234729.2 | XP_006234791.1 | 14 | 1787 | |||
XM_008762080.1 | XP_008760302.1 | 14 | 2691 | |||
XM_008762081.1 | XP_008760303.1 | 14 | 2691 | |||
GenBank mRNA | AF052540.1 | - | 13 | 1567 | ||
AF061726.2 | - | 14 | 1764 | |||
AF184950.1 | - | 14 | 1704 | |||
J05121.1 | - | 14 | 2059 | |||
U96367.1 | - | 13 | 1616 |
